Learn More
Cellular Dynamics International iCell Cardiomyocytes, 01434, 4 x 10^6 cells/vial
Catalog No. NC1431328 Shop All Cellular Dynamics International Products
Encompass
Heart disease encompasses a wide range of conditions that can be a result of genetics, physiologic, and metabolic disorders as well as adverse drug reactions. The availability of human cell models that could be used to interrogate these various factors would have a profound impact on the effort to find new medicines and cures for heart disease. Derived from induced pluripotent stem cells (iPSCs), iCell Cardiomyocytes from FUJIFILM Cellular Dynamics, Inc. (FCDI), enable a wide range of applications spanning disease research, drug discovery, safety and toxicity testing, and regenerative medicine.
Catalog No. NC1431328 Supplier Cellular Dynamics International Supplier No. R1007
Heart disease encompasses a wide range of conditions that can be a result of genetics, physiologic, and metabolic disorders as well as adverse drug reactions. The availability of human cell models that could be used to interrogate these various factors would have a profound impact on the effort to find new medicines and cures for heart disease. Derived from induced pluripotent stem cells (iPSCs), iCell Cardiomyocytes from FUJIFILM Cellular Dynamics, Inc. (FCDI), enable a wide range of applications spanning disease research, drug discovery, safety and toxicity testing, and regenerative medicine.