Learn More
Cellular Dynamics International iCell Cardiomyocytes, cryopreserved, 4 x 10^6 viable cells per vial, >95% purity, human iPSC-derived, Donor ID 11713
Catalog No. NC1677665 Shop All Cellular Dynamics International Products
Encompass
The FUJIFILM iCell Cardiomyocytes are a premier choice for in vitro cardiac research, derived from human induced pluripotent stem cells (iPSCs). This product offers a rich source of cardiomyocytes suitable for various applications, including drug discovery and toxicity testing. Each vial contains a high concentration of viable cells that exhibit essential cardiac functionalities, such as spontaneous beating and proper calcium handling, making them a gold standard in the field.
  • High purity greater than 95% cardiomyocytes
  • 4 x 10^6 viable cells per vial
  • Cryopreserved format for convenience
  • Compatible with optimized media for maximum performance
  • Utilized in drug discovery and disease modeling
  • Features spontaneous beating and intact excitation-contraction coupling
  • Recommended for culture duration of up to 2 weeks
Catalog No. NC1677665 Supplier Cellular Dynamics International Supplier No. R1106
The FUJIFILM iCell Cardiomyocytes are a premier choice for in vitro cardiac research, derived from human induced pluripotent stem cells (iPSCs). This product offers a rich source of cardiomyocytes suitable for various applications, including drug discovery and toxicity testing. Each vial contains a high concentration of viable cells that exhibit essential cardiac functionalities, such as spontaneous beating and proper calcium handling, making them a gold standard in the field.
  • High purity greater than 95% cardiomyocytes
  • 4 x 10^6 viable cells per vial
  • Cryopreserved format for convenience
  • Compatible with optimized media for maximum performance
  • Utilized in drug discovery and disease modeling
  • Features spontaneous beating and intact excitation-contraction coupling
  • Recommended for culture duration of up to 2 weeks