Learn More
Cellular Dynamics International iCell Cardiomyocytes Kit 3 Pack, 01434
Catalog No. NC1650716 Shop All Cellular Dynamics International Products
Encompass
Each kit contains: 3 Vials Cryopreserved iCell Cardiomyocytes, 01434, 30 ml Plating Medium, 2 x 100 ml Maintenance Medium, User’s Guide. 3 Vials at ≥1 x 106 Viable Cells/Vial
Catalog No. NC1650716 Supplier Cellular Dynamics International Supplier No. R1132
Each kit contains: 3 Vials Cryopreserved iCell Cardiomyocytes, 01434, 30 ml Plating Medium, 2 x 100 ml Maintenance Medium, User’s Guide. 3 Vials at ≥1 x 106 Viable Cells/Vial