Learn More
FUJIFILM BIOSCIENCES INC iCell DopaNeurons, 01279, 1 x 10^6 cells/vial
Supplier:  FUJIFILM BIOSCIENCES INC R1088
Encompass
iCell DopaNeurons are iPS cell-derived human midbrain floorplate dopaminergic neurons, differentiated in protocols licensed and adapted from the Lorenz Studer lab (Memorial Sloan Kettering) and industrialized for scale at FCDI.
Catalog No. NC1867936