Learn More
Cellular Dynamics International iCell DopaNeurons 01279, Human iPSC-derived midbrain dopaminergic neurons, >70% TH+, cryopreserved
Catalog No. NC1999236 Shop All Cellular Dynamics International Products
Encompass
iCell DopaNeurons 01279 are a highly pure, fully differentiated population of human induced pluripotent stem cell (iPSC)-derived midbrain dopaminergic neurons. They are suitable for targeted drug discovery, neurotoxicity testing, and studies in regenerative medicine, particularly for Parkinson's disease. This product is designed to provide a biologically relevant in vitro model for neurological research, exhibiting key physiological characteristics like spontaneous and evoked action potentials, as well as functional dopamine release and uptake.
  • Derived from human iPSCs to ensure physiological relevance.
  • Consistently manufactured with a purity of >80% TH+ neurons.
  • Exhibit spontaneous and evoked action potentials and dopamine functionality.
  • Shipped cryopreserved with optimized media for ease of use.
  • Compatible with various biochemical and cell-based assays.
Catalog No. NC1999236 Supplier Cellular Dynamics International Supplier No. C1028
Add to Cart
Add to Cart
iCell DopaNeurons 01279 are a highly pure, fully differentiated population of human induced pluripotent stem cell (iPSC)-derived midbrain dopaminergic neurons. They are suitable for targeted drug discovery, neurotoxicity testing, and studies in regenerative medicine, particularly for Parkinson's disease. This product is designed to provide a biologically relevant in vitro model for neurological research, exhibiting key physiological characteristics like spontaneous and evoked action potentials, as well as functional dopamine release and uptake.
  • Derived from human iPSCs to ensure physiological relevance.
  • Consistently manufactured with a purity of >80% TH+ neurons.
  • Exhibit spontaneous and evoked action potentials and dopamine functionality.
  • Shipped cryopreserved with optimized media for ease of use.
  • Compatible with various biochemical and cell-based assays.