Learn More
Cellular Dynamics International iCell DopaNeurons, Cryopreserved, 3 vials, > 1 x 10^6 cells/vial, Fully differentiated, Human, Midbrain floorplate, >80% TH+, ≥90% MAP2+, ≥50% FoxA2+, ≥50% TH+, Branched networks ≤ 3 days
Catalog No. NC2598496 Shop All Cellular Dynamics International Products
Encompass
iCell DopaNeurons, Cryopreserved, 3 vials, > 1 x 10^6 cells/vial, Fully differentiated, Human, Midbrain floorplate, >80% TH+, ≥90% MAP2+, ≥50% FoxA2+, ≥50% TH+, Branched networks ≤ 3 days
Catalog No. NC2598496 Supplier Cellular Dynamics International Supplier No. R1108
iCell DopaNeurons, Cryopreserved, 3 vials, > 1 x 10^6 cells/vial, Fully differentiated, Human, Midbrain floorplate, >80% TH+, ≥90% MAP2+, ≥50% FoxA2+, ≥50% TH+, Branched networks ≤ 3 days