Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellular Dynamics International iCell DopaNeurons PD GBA N370S Kit, 5.0 x 10^6 viable cells, cryopreserved kit, >80% TH-positive neurons, fully differentiated neurons, includes media supplements

Supplier:  Cellular Dynamics International R1231

Encompass

The iCell DopaNeurons PD GBA N370S Kit from FUJIFILM CELLULAR DYNAMICS INC provides cryopreserved, human iPSC-derived midbrain dopaminergic neurons specifically designed for Parkinson's disease research. These cells are tailored to model the GBA N370S mutation, offering a biologically relevant in vitro system ideal for therapeutic discovery and drug screening. They are derived from participants in the Michael J. Fox Foundation's PPMI study, ensuring their physiological relevance through comprehensive clinical and biological data.

  • Cell type: Human iPSC-derived midbrain dopaminergic neurons
  • Disease model: Parkinson's disease
  • Genetic mutation: GBA (N370S)
  • Purity: >80% tyrosine hydroxylase (TH)-positive
  • Fully differentiated neurons
  • Quantity: >= 5.0 x 10^6 viable cells
  • Optimal media included for various assays
  • Functional recovery within 14 days of plating
  • Shipping condition: Cryopreserved cells

Catalog No. NC3232480


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.