Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellular Dynamics International iCell Endothelial Cells 01434, Catalog #C1021, Cryopreserved, ≥ 1 x 10^6 cells/vial

Catalog No. NC2373845 Shop All Cellular Dynamics International Products
Encompass

FUJIFILM Cellular Dynamics iCell Endothelial Cells 01434 are highly pure, human iPSC-derived endothelial cells ideal for vascular research. These cryopreserved cells ensure reliable and physiologically relevant models for studying vascular diseases, drug discovery, and tissue engineering.

  • Exhibit consistent expression of endothelial surface markers
  • Demonstrate characteristic functional behaviors for reproducibility in modeling
  • Suitable for diverse applications in monoculture and co-culture systems
  • Purity greater than 90% CD31+/CD105+
  • Viable in culture for weeks
  • Quantity: ≥ 1 x 10⁶ cells/vial
  • Storage conditions: -150℃ or liquid nitrogen

Catalog No. NC2373845 Supplier Cellular Dynamics International Supplier No. C1021
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FUJIFILM Cellular Dynamics iCell Endothelial Cells 01434 are highly pure, human iPSC-derived endothelial cells ideal for vascular research. These cryopreserved cells ensure reliable and physiologically relevant models for studying vascular diseases, drug discovery, and tissue engineering.

  • Exhibit consistent expression of endothelial surface markers
  • Demonstrate characteristic functional behaviors for reproducibility in modeling
  • Suitable for diverse applications in monoculture and co-culture systems
  • Purity greater than 90% CD31+/CD105+
  • Viable in culture for weeks
  • Quantity: ≥ 1 x 10⁶ cells/vial
  • Storage conditions: -150℃ or liquid nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.