Learn More
Cellular Dynamics International iCell GABANeurons, 01279, 4 x 10^6 cells/vial
Catalog No. NC1808433 Shop All Cellular Dynamics International Products
Encompass
iCell GABANeurons are iPS cell-derived mixed populations of human cerebral cortical neurons that exhibit native electrical and biochemical activity. They overcome limitations of existing models.
Catalog No. NC1808433 Supplier Cellular Dynamics International Supplier No. R1011
iCell GABANeurons are iPS cell-derived mixed populations of human cerebral cortical neurons that exhibit native electrical and biochemical activity. They overcome limitations of existing models.