Learn More
Cellular Dynamics International iCell GlutaNeurons, Human glutamatergic cortical neurons, ≥90% purity, ≥6.0 x 10^6 viable cells/vial
Catalog No. NC1999237 Shop All Cellular Dynamics International Products
Encompass
FUJIFILM IRVINE SCIENTIFIC iCell GlutaNeurons are human glutamatergic-enriched cortical neurons derived from induced pluripotent stem cells. These neurons form functional neuronal networks that are highly relevant to neurological research, providing a valuable tool for various scientific applications including drug discovery and toxicity testing.
  • Biologically relevant model for human neurological research
  • Self-assembling into active excitatory neural networks
  • Pure composition of ≥90% glutamatergic neurons
  • Suitable for diverse applications including investigating neurotoxicity and epilepsy
  • Cells have extended viability, remaining viable for >4 weeks
  • Compatible with various assay techniques including electrophysiology and calcium signaling
  • Ideal for modeling neurodegenerative conditions
Catalog No. NC1999237 Supplier Cellular Dynamics International Supplier No. C1033
FUJIFILM IRVINE SCIENTIFIC iCell GlutaNeurons are human glutamatergic-enriched cortical neurons derived from induced pluripotent stem cells. These neurons form functional neuronal networks that are highly relevant to neurological research, providing a valuable tool for various scientific applications including drug discovery and toxicity testing.
  • Biologically relevant model for human neurological research
  • Self-assembling into active excitatory neural networks
  • Pure composition of ≥90% glutamatergic neurons
  • Suitable for diverse applications including investigating neurotoxicity and epilepsy
  • Cells have extended viability, remaining viable for >4 weeks
  • Compatible with various assay techniques including electrophysiology and calcium signaling
  • Ideal for modeling neurodegenerative conditions