Learn More
Cellular Dynamics International iCELL MACROPHAGES 2.0 Kit, Macrophage cell type, human, ≥ 1 x 10^6 viable cells, >90% CD68+ purity, up to 14 days culture duration
Catalog No. NC2734771 Shop All Cellular Dynamics International Products
Encompass
iCELL MACROPHAGES 2.0 Kit, Macrophage cell type, human, ≥ 1 x 10^6 viable cells, >90% CD68+ purity, up to 14 days culture duration
Catalog No. NC2734771 Supplier Cellular Dynamics International Supplier No. R1186
iCELL MACROPHAGES 2.0 Kit, Macrophage cell type, human, ≥ 1 x 10^6 viable cells, >90% CD68+ purity, up to 14 days culture duration