Learn More
Cellular Dynamics International iCell Microglia Kit, AD APOE E4/E4, 1 x 10^6 cells/vial, Cryopreserved, fully differentiated, Research Use Only
Catalog No. NC3761803 Shop All Cellular Dynamics International Products
Encompass
The iCell Microglia Kit is designed for advanced Alzheimer's Disease (AD) research, providing consistent and reproducible human induced pluripotent stem cell (iPSC)-derived microglia. These cryopreserved, fully differentiated cells overcome the limitations of primary human microglia for studying neurodegenerative diseases. Utilized in a variety of applications, this kit includes components for optimal cell maintenance and functionality, enabling researchers to delve into cytokine signaling and synaptic transmission.
  • iPSC-derived microglia
  • Specifically tailored for Alzheimer's Disease studies
  • Genotype: APOE E4/E4
  • Quantity: ≥ 1 x 10^6 cells/vial
  • Cryopreserved and fully differentiated cell state
  • Supports research use only
  • Includes complete maintenance medium and supplements
  • Maintains physiological functions including phagocytosis and synaptic plasticity regulation
Catalog No. NC3761803 Supplier Cellular Dynamics International Supplier No. R1213
Add to Cart
Edge
Add to Cart
The iCell Microglia Kit is designed for advanced Alzheimer's Disease (AD) research, providing consistent and reproducible human induced pluripotent stem cell (iPSC)-derived microglia. These cryopreserved, fully differentiated cells overcome the limitations of primary human microglia for studying neurodegenerative diseases. Utilized in a variety of applications, this kit includes components for optimal cell maintenance and functionality, enabling researchers to delve into cytokine signaling and synaptic transmission.
  • iPSC-derived microglia
  • Specifically tailored for Alzheimer's Disease studies
  • Genotype: APOE E4/E4
  • Quantity: ≥ 1 x 10^6 cells/vial
  • Cryopreserved and fully differentiated cell state
  • Supports research use only
  • Includes complete maintenance medium and supplements
  • Maintains physiological functions including phagocytosis and synaptic plasticity regulation