Learn More
Cellular Dynamics International iCell Microglia Media Kit, 50 ml iCell Glial Base Medium, 0.5 ml iCell Microglia Supplement A (100x), 0.5 ml iCell Microglia Supplement B (100x), 1 ml iCell Neural Supplement C (50X)
Catalog No. NC3363787 Shop All Cellular Dynamics International Products
Encompass
The iCell Microglia Media Kit is a specialized cell culture medium tailored for the optimal growth and maintenance of human iPSC-derived microglia. It provides a consistent in vitro environment, essential for research in microglial biology and neurodegenerative diseases. This media kit solves the challenges faced with primary human microglia by ensuring reproducible results for various experiments related to neuroinflammation and drug discovery.
  • Supports growth of iPSC-derived human microglia.
  • Includes multiple supplements for enhanced performance.
  • Stores complete medium at 4°C for up to 2 weeks.
  • Can be aliquoted for single-use storage at -20°C for up to 3 months.
  • Recommended culture conditions: 37°C and 5% CO2.
  • Designed for research purposes only, focusing on neurodegeneration and drug discovery.
  • Maintains relevant microglial markers and physiological functions.
Catalog No. NC3363787 Supplier Cellular Dynamics International Supplier No. R1204
The iCell Microglia Media Kit is a specialized cell culture medium tailored for the optimal growth and maintenance of human iPSC-derived microglia. It provides a consistent in vitro environment, essential for research in microglial biology and neurodegenerative diseases. This media kit solves the challenges faced with primary human microglia by ensuring reproducible results for various experiments related to neuroinflammation and drug discovery.
  • Supports growth of iPSC-derived human microglia.
  • Includes multiple supplements for enhanced performance.
  • Stores complete medium at 4°C for up to 2 weeks.
  • Can be aliquoted for single-use storage at -20°C for up to 3 months.
  • Recommended culture conditions: 37°C and 5% CO2.
  • Designed for research purposes only, focusing on neurodegeneration and drug discovery.
  • Maintains relevant microglial markers and physiological functions.