Learn More
Cellular Dynamics International iCell Motor Neurons Kit, R1051, 01279, human iPSC-derived spinal motor neurons, ≥1.0 x 10^6 viable cells per vial, cryopreserved, includes base medium 100 ml, supplement A 2 ml, nervous system supplement 1 ml
Catalog No. NC2373849 Shop All Cellular Dynamics International Products
Encompass
iCell Motor Neurons Kit, R1051, 01279, human iPSC-derived spinal motor neurons, ≥1.0 x 10^6 viable cells per vial, cryopreserved, includes base medium 100 ml, supplement A 2 ml, nervous system supplement 1 ml
Catalog No. NC2373849 Supplier Cellular Dynamics International Supplier No. R1051
iCell Motor Neurons Kit, R1051, 01279, human iPSC-derived spinal motor neurons, ≥1.0 x 10^6 viable cells per vial, cryopreserved, includes base medium 100 ml, supplement A 2 ml, nervous system supplement 1 ml