Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellular Dynamics International iCell Motor Neurons Kit 01279, Human iPSC-derived spinal motor neurons, ≥90% β-III tubulin+, ≥70% ISL1/2+, Viability ≥14 days, 3 x 10^6 cells/vial

Catalog No. NC2373850 Shop All Cellular Dynamics International Products
Encompass

iCell Motor Neurons Kit 01279 is a highly pure, functional population of human spinal motor neurons derived from induced pluripotent stem cells (iPSCs) by FUJIFILM Cellular Dynamics Inc. This kit offers a reliable source of human motor neurons with typical morphology, strong viability in culture, and consistent gene expression. These cells are an essential resource for scientific applications, including neurodegenerative disease research, drug development, and toxicology screening.

  • Human iPSC-derived spinal motor neurons
  • Purity ≥90% β-III tubulin+, ≥70% ISL1/2+ at Day 14
  • Viability in culture for ≥14 days
  • Typical motor neuron morphology with branching neurites
  • Key markers expressed: SMI32
  • Applications include neurodegenerative disease research, drug discovery, and bioengineering
  • Quantity: 3 x 10^6 cells/vial

Catalog No. NC2373850 Supplier Cellular Dynamics International Supplier No. R1049
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

iCell Motor Neurons Kit 01279 is a highly pure, functional population of human spinal motor neurons derived from induced pluripotent stem cells (iPSCs) by FUJIFILM Cellular Dynamics Inc. This kit offers a reliable source of human motor neurons with typical morphology, strong viability in culture, and consistent gene expression. These cells are an essential resource for scientific applications, including neurodegenerative disease research, drug development, and toxicology screening.

  • Human iPSC-derived spinal motor neurons
  • Purity ≥90% β-III tubulin+, ≥70% ISL1/2+ at Day 14
  • Viability in culture for ≥14 days
  • Typical motor neuron morphology with branching neurites
  • Key markers expressed: SMI32
  • Applications include neurodegenerative disease research, drug discovery, and bioengineering
  • Quantity: 3 x 10^6 cells/vial

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.