Learn More
Cellular Dynamics International iCell Neural Supplement C, 1 ml, Liquid concentrate, 50X, Research Use Only, for iPSC-derived neural cell culture
Catalog No. NC2947946 Shop All Cellular Dynamics International Products
Encompass
FUJIFILM Cellular Dynamics Inc. iCell Neural Supplement C is a serum-free supplement intended for human induced pluripotent stem cell (iPSC)-derived neural cell culture. This product enhances the maintenance of various neural cell types and is essential for advanced research applications such as drug discovery and neurotoxicity studies. The product's formulation supports optimal growth and function, providing a vital resource for neuroscience research.
Catalog No. NC2947946 Supplier Cellular Dynamics International Supplier No. M1055
FUJIFILM Cellular Dynamics Inc. iCell Neural Supplement C is a serum-free supplement intended for human induced pluripotent stem cell (iPSC)-derived neural cell culture. This product enhances the maintenance of various neural cell types and is essential for advanced research applications such as drug discovery and neurotoxicity studies. The product's formulation supports optimal growth and function, providing a vital resource for neuroscience research.