Learn More
Cellular Dynamics International iCell Neural Supplement B, Serum-free, 2 ml
Catalog No. NC2883854 Shop All Cellular Dynamics International Products
Encompass
iCell Neural Supplement B is a serum-free medium supplement specifically designed to support the health and function of iPSC-derived neural cell types, such as dopaminergic and glutamatergic neurons. This supplement is crucial for maintaining long-term viability while limiting the proliferation of progenitor or non-neuronal cells in culture. Shipped frozen and requiring storage at -20°C, it is ideal for various neuroscience applications, including disease modeling and drug discovery for neurodegenerative conditions like Parkinson's disease.
  • Supports neuronal health and function
  • Limits progenitor cell proliferation
  • Suitable for dopaminergic and glutamatergic neurons
  • Serum-free formulation
  • Shipped frozen on dry ice
  • Optimal storage at -20°C
  • Part of complete maintenance medium
  • Antibiotic-free
  • Useful for Parkinson's disease research
  • Facilitates drug discovery applications
Catalog No. NC2883854 Supplier Cellular Dynamics International Supplier No. M1029
iCell Neural Supplement B is a serum-free medium supplement specifically designed to support the health and function of iPSC-derived neural cell types, such as dopaminergic and glutamatergic neurons. This supplement is crucial for maintaining long-term viability while limiting the proliferation of progenitor or non-neuronal cells in culture. Shipped frozen and requiring storage at -20°C, it is ideal for various neuroscience applications, including disease modeling and drug discovery for neurodegenerative conditions like Parkinson's disease.
  • Supports neuronal health and function
  • Limits progenitor cell proliferation
  • Suitable for dopaminergic and glutamatergic neurons
  • Serum-free formulation
  • Shipped frozen on dry ice
  • Optimal storage at -20°C
  • Part of complete maintenance medium
  • Antibiotic-free
  • Useful for Parkinson's disease research
  • Facilitates drug discovery applications