Learn More
Cellular Dynamics International iCell Sensory Neurons 01279 Kit, R1251, ≥ 90% purity, ≥ 1.0 x 10^6 viable cells, cryopreserved single-cell suspension, human-derived
Catalog No. NC3368094 Shop All Cellular Dynamics International Products
Encompass
iCell Sensory Neurons 01279 Kit, R1251, ≥ 90% purity, ≥ 1.0 x 10^6 viable cells, cryopreserved single-cell suspension, human-derived
Catalog No. NC3368094 Supplier Cellular Dynamics International Supplier No. R1251
iCell Sensory Neurons 01279 Kit, R1251, ≥ 90% purity, ≥ 1.0 x 10^6 viable cells, cryopreserved single-cell suspension, human-derived