Learn More
Alkali Scientific IceRack™ for 72 × Tubes, detachable lid, tube platform, and base, numbered cavities, snap cap and screw cap compatibile, 6/cs, 100 × 1.5mL tubes
SDP
Catalog No. 5000662282
Encompass_Preferred
Crushed Ice Microtube Workstations These new IceRack™ tube boxes have a removable white tube tray that can be kept filled with crushed ice or other temperature controlling materials. They also have a translucent cover. Temperature sensitive samples and assays in microtubes can be processed with an extra degree of temperature stability. Suitable for use with screw cap or snap cap microtubes from 0.5mL to 1.5/2.0mL Supplied 6 boxes per pack. Designed for crushed Ice Polypropylene, 80°C to 135°C Detachable “lift off” lids Economical For screw cap and snap cap tubes
Catalog No. 50-006-62282 Supplier Alkali Scientific Supplier No. AR3814
Crushed Ice Microtube Workstations These new IceRack™ tube boxes have a removable white tube tray that can be kept filled with crushed ice or other temperature controlling materials. They also have a translucent cover. Temperature sensitive samples and assays in microtubes can be processed with an extra degree of temperature stability. Suitable for use with screw cap or snap cap microtubes from 0.5mL to 1.5/2.0mL Supplied 6 boxes per pack. Designed for crushed Ice Polypropylene, 80°C to 135°C Detachable “lift off” lids Economical For screw cap and snap cap tubes