Learn More
Research Products International Corp IceRack Tube Workstation, 72 x 0.5 ml, 6/pk
SDP
Catalog No. 502252999
Encompass
IceRack Tube Workstation features removable white tube tray that holds micro-tubes and crushed ice. Ideal for use with temperature sensitive samples and assays that require additional cold temperature stability on a benchtop. Suitable for use with screw cap or snap cap microtubes (0.5 to 1.5/2.0ml, see ordering options) and withstand temperatures from -80°C to 135°C. Economical design features detachable translucent “lift-off” lid that allows full access of contents. Supplied 6 boxes per pack.
Holds crushed ice and micro-tubes
Detachable lift-off lid
Polyproylene
This item is not returnable. View return policy
Catalog No. 50-225-2999 Supplier Research Products International Corp Supplier No. 248525
IceRack Tube Workstation features removable white tube tray that holds micro-tubes and crushed ice. Ideal for use with temperature sensitive samples and assays that require additional cold temperature stability on a benchtop. Suitable for use with screw cap or snap cap microtubes (0.5 to 1.5/2.0ml, see ordering options) and withstand temperatures from -80°C to 135°C. Economical design features detachable translucent “lift-off” lid that allows full access of contents. Supplied 6 boxes per pack.
Holds crushed ice and micro-tubes
Detachable lift-off lid
Polyproylene