Learn More
Medchemexpress LLC Icerguastat (Sephin1) | 951441-04-6 | 99.93% | 196.64 | 10 MG
SDP
Catalog No. 5000343618
Encompass_Preferred
Icerguastat (Sephin1) is a Guanabenz derivative that selectively inhibits the phosphatase regulatory subunit PPP1R15A (R15A). It functions by inhibiting eIF2α dephosphorylation, which prolongs the protective cellular response and demonstrates an anti-prion effect.
  • Selectively inhibits the phosphatase regulatory subunit PPP1R15A (R15A).
  • Inhibits eIF2α dephosphorylation, extending protective cellular responses.
  • Demonstrates anti-prion effects in models.
  • Prolongs eIF2α phosphorylation in stressed oligodendrocytes.
  • Delays the onset of experimental autoimmune encephalomyelitis (EAE) in animal models.
  • Prolongs survival in prion-infected mice.
Catalog No. 50-003-43618 Supplier Medchemexpress LLC Supplier No. HY111022100MG
Icerguastat (Sephin1) is a Guanabenz derivative that selectively inhibits the phosphatase regulatory subunit PPP1R15A (R15A). It functions by inhibiting eIF2α dephosphorylation, which prolongs the protective cellular response and demonstrates an anti-prion effect.
  • Selectively inhibits the phosphatase regulatory subunit PPP1R15A (R15A).
  • Inhibits eIF2α dephosphorylation, extending protective cellular responses.
  • Demonstrates anti-prion effects in models.
  • Prolongs eIF2α phosphorylation in stressed oligodendrocytes.
  • Delays the onset of experimental autoimmune encephalomyelitis (EAE) in animal models.
  • Prolongs survival in prion-infected mice.