Learn More
Medchemexpress LLC HY-14428 10mg Medchemexpress, ICG-001 CAS:780757-88-2 Purity:>98%
SDP
Catalog No. 502029049
Encompass_Preferred
Medchemexpress, HY-14428 10mg ICG-001 CAS:780757-88-2 ICG-001 is an inhibitor of β-catenin/TCF mediated transcription. ICG-001 works by specifically binding to cyclic AMP response element-binding protein with an IC50 of 3 μM. ICG-001 selectively blocks the β-catenin/CBP interaction without interfering with the β-catenin/p300 interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9049 Supplier Medchemexpress LLC Supplier No. HY1442810MG
Medchemexpress, HY-14428 10mg ICG-001 CAS:780757-88-2 ICG-001 is an inhibitor of β-catenin/TCF mediated transcription. ICG-001 works by specifically binding to cyclic AMP response element-binding protein with an IC50 of 3 μM. ICG-001 selectively blocks the β-catenin/CBP interaction without interfering with the β-catenin/p300 interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.