Learn More
Apexbio Technology LLC ICG 001 847591-62-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062103
Encompass_Preferred
ICG 001 is a small molecule inhibitor of the canonical Wnt/ -catenin signaling pathway Mechanistically it competes with -catenin for interaction with CREB-binding protein (CBP) thereby selectively disrupting CBP/ -catenin-mediated transcriptional activity driven by the TCF/ -catenin complex (IC50 3 M) Experimental studies demonstrated its use in assessing CBP-dependent biological functions regulated by Wnt signaling Research applications include investigating CBP/ -catenin roles in cancer cell proliferation such as colorectal cancer cells (HCT-116 SW480) glioblastoma stem cell growth and fibrotic responses in pulmonary and dermal experimental models Additionally ICG 001 is under evaluation in clinical studies aiming at treatment of colorectal cancer and leukemia
Catalog No. 50-000-62103 Supplier Apexbio Technology LLC Supplier No. A82175.1
ICG 001 is a small molecule inhibitor of the canonical Wnt/ -catenin signaling pathway Mechanistically it competes with -catenin for interaction with CREB-binding protein (CBP) thereby selectively disrupting CBP/ -catenin-mediated transcriptional activity driven by the TCF/ -catenin complex (IC50 3 M) Experimental studies demonstrated its use in assessing CBP-dependent biological functions regulated by Wnt signaling Research applications include investigating CBP/ -catenin roles in cancer cell proliferation such as colorectal cancer cells (HCT-116 SW480) glioblastoma stem cell growth and fibrotic responses in pulmonary and dermal experimental models Additionally ICG 001 is under evaluation in clinical studies aiming at treatment of colorectal cancer and leukemia