Learn More
Apexbio Technology LLC ICG 001, 25mg. Cas: 847591-62-2 MFCD: MFCD12196918. Wnt/β-catenin pathway inhibitor
SDP
Supplier:  Apexbio Technology LLC A821725
Encompass_Preferred
ICG001 is a small molecule inhibitor that targets Wnt/-catenin pathway. It inhibits TCF/-catenin mediated transcription by competing with -catenin for CBP (CREB binding protein) but not p300 binding with an IC50 of 3 M [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5078