Learn More
Selleck Chemical LLC ICG-001 5mg 780757-88-2 (relative stereochemistry); 847591-62-2 (absolute stereochemistry)
SDP
Supplier:  Selleck Chemical LLC S26625MG
Encompass_Preferred
ICG-001 antagonizes Wnt/?-catenin/TCF-mediated transcription and specifically binds to CREB-binding protein (CBP) with IC50 of 3 ?M, but is not the related transcriptional coactivator p300. ICG-001 induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7949