Learn More
Medchemexpress LLC Icg-amine | 1686147-55-6 | 98.3% | 829.14 g/mol | C51H64N4O4S | 5 MG
SDP
Catalog No. 5000263092
Encompass_Preferred
ICG-amine is a near-infrared tricarbocyanine fluorescent probe that reacts with amino acid residues without requiring condensing agents. Supplied as a green to dark green solid, it is used for in vivo and animal imaging applications, including cancer imaging and studies of metabolism and metastasis.
  • Binds amino acid residues without condensing agents.
  • Near-infrared fluorescence suitable for deep tissue imaging.
  • High purity (98.34%) for research use.
  • Soluble in DMSO and water with heat and sonication; compatible with common in vivo formulations.
  • Provided as a solid with a defined molecular weight and molecular formula for characterization.
Catalog No. 50-002-63092 Supplier Medchemexpress LLC Supplier No. HY1338845MG
ICG-amine is a near-infrared tricarbocyanine fluorescent probe that reacts with amino acid residues without requiring condensing agents. Supplied as a green to dark green solid, it is used for in vivo and animal imaging applications, including cancer imaging and studies of metabolism and metastasis.
  • Binds amino acid residues without condensing agents.
  • Near-infrared fluorescence suitable for deep tissue imaging.
  • High purity (98.34%) for research use.
  • Soluble in DMSO and water with heat and sonication; compatible with common in vivo formulations.
  • Provided as a solid with a defined molecular weight and molecular formula for characterization.