Learn More
Medchemexpress LLC Icg-amine | 1686147-55-6 | 98.3% | C51H64N4O4S | 10MG
SDP
Catalog No. 5000259939
Encompass_Preferred
ICG-amine is a near-infrared fluorescent probe derived from indocyanine green that binds to amino acid residues without the need for condensing agents. It is intended for fluorescence labeling and imaging applications that require NIR properties and includes recommended solubility and storage instructions for research use.
  • Provides near-infrared fluorescence useful for deep-tissue imaging.
  • Binds to amino acid residues without requiring condensing agents.
  • Reported high purity supports research-grade applications.
  • Soluble in DMSO and water with sonication or gentle heating as needed.
  • Powder form stable at -20°C with extended shelf life when stored appropriately.
Catalog No. 50-002-59939 Supplier Medchemexpress LLC Supplier No. HY13388410MG
ICG-amine is a near-infrared fluorescent probe derived from indocyanine green that binds to amino acid residues without the need for condensing agents. It is intended for fluorescence labeling and imaging applications that require NIR properties and includes recommended solubility and storage instructions for research use.
  • Provides near-infrared fluorescence useful for deep-tissue imaging.
  • Binds to amino acid residues without requiring condensing agents.
  • Reported high purity supports research-grade applications.
  • Soluble in DMSO and water with sonication or gentle heating as needed.
  • Powder form stable at -20°C with extended shelf life when stored appropriately.