Learn More
Medchemexpress LLC HY-D1041 5mg , ICG-OSu CAS:1622335-40-3 Purity:>98%
SDP
Catalog No. 502261937
Encompass_Preferred
HY-D1041 5mg ICG-OSu CAS: 1622335-40-3 ICG-OSu (ICG NHS ester), a near-infrared fluorescent agent, is amine-reactive and has been widely used to design in vivo imaging probes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1937 Supplier Medchemexpress LLC Supplier No. HYD10415MG
HY-D1041 5mg ICG-OSu CAS: 1622335-40-3 ICG-OSu (ICG NHS ester), a near-infrared fluorescent agent, is amine-reactive and has been widely used to design in vivo imaging probes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.