Learn More
Medchemexpress LLC Icg-sulfo-osu sodium | 190714-28-4 | MFCD11045310 | 90.3% | C49H52N3NaO10S2 | 10MG
SDP
Catalog No. 5000259229
Encompass_Preferred
ICG-Sulfo-OSu sodium is a PEG-based linker bearing an indocyanine green-derived sulfo-N-hydroxysuccinimide (sulfo-OSu) ester. Supplied as the sodium salt in solid form, it provides an activated amine-reactive handle for conjugation and is intended for research use in assembling PROTACs and other bioconjugates. Store sealed, away from moisture and light.
Catalog No. 50-002-59229 Supplier Medchemexpress LLC Supplier No. HY13471910MG
ICG-Sulfo-OSu sodium is a PEG-based linker bearing an indocyanine green-derived sulfo-N-hydroxysuccinimide (sulfo-OSu) ester. Supplied as the sodium salt in solid form, it provides an activated amine-reactive handle for conjugation and is intended for research use in assembling PROTACs and other bioconjugates. Store sealed, away from moisture and light.