Learn More
Aat Bioquest ICG-Sulfo-OSu
SDP
Catalog No. 501952790
Encompass_Preferred
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics. It is used for determining cardiac output, hepatic function, and liver blood flow, and for ophthalmic angiography. It has a peak spectral absorption close to 800 nm.
Catalog No. 50-195-2790 Supplier Aat Bioquest Supplier No. 180
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics. It is used for determining cardiac output, hepatic function, and liver blood flow, and for ophthalmic angiography. It has a peak spectral absorption close to 800 nm.