Learn More
TARGETMOL CHEMICALS INC ICG001 10MG
SDP
Catalog No. 502577190
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ICG001 (PRI-724 isomer) antagonizes Wnt/(beta)-catenin/TCF-mediated transcription and specifically binds to element-binding protein (CREB)-binding protein (CBP) with IC50 of 3 uM but is not the related transcriptional coactivator p300. purity: 99%
Catalog No. 50-257-7190 Supplier TARGETMOL CHEMICALS INC Supplier No. T223710MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ICG001 (PRI-724 isomer) antagonizes Wnt/(beta)-catenin/TCF-mediated transcription and specifically binds to element-binding protein (CREB)-binding protein (CBP) with IC50 of 3 uM but is not the related transcriptional coactivator p300. purity: 99%