Learn More
BINDER™ ICH Light Module
SDP
Catalog No. 502875585 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB PRO 720, KBF/KBF PRO 720
KB PRO/KBF/KBF PRO 260/470
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5585 KB PRO 720, KBF/KBF PRO 720
50-287-5584 KB PRO/KBF/KBF PRO 260/470
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-287-5585 Supplier BINDER™ Supplier No. 80122442
Add to Cart
Add to Cart
Request a Quote
ICH-Q1B light module
Module consists of:
  • VIS cassette
  • UVA cassette
  • Independent power supply
Specifications
For Use With (Equipment) KB PRO 720, KBF/KBF PRO 720
Material Glass
Type Lighting