Learn More
Darwin Chambers Company Triple Door ICH Stability Chamber 66 cu. ft.
SDP
Supplier:  Darwin Chambers Company PH084SS
Encompass_Preferred
The PH084-SS is a 76.2 cubic ft triple door stability chamber. These Chambers are designed for performance specifications exceeding FDA & ICH requirements. Temperature range is 20°C to 50°C degrees C. Humidity range ambient to 90 percent. This unit comes w/ nine pvc coated shelves. The chamber interior provides stainless steel side walls & back, stainless steel floor & ceiling. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. The 7 year warranty on cooling components & the ability of our chambers to maintain temperature even if the chamber suffers multiple failures is unmatched by any other refrigeration-based chamber. The ultrasonic humidification system provides excellent humidity control & avoids hot spots seen during chamber mapping of steam boiler equipped chambers. The PH Series chambers utilize less energy than comparable stability chambers.
Catalog No. 50-189-3935
Add to Cart
Request a Quote