Learn More
Bioworld Ichihashi New Phalaenopsis (NP) Medium, 50 L
Supplier:  Bioworld 306301963
Encompass_Preferred
Ichihashi New Phalaenopsis (NP) MediumContains the Macronutrients and Micronutrients as described by Ichihashi (1992) Modified to contain NH4NO3Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 25.35 g/LSoluble in water
Catalog No. 50-254-990