Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ichthammol, USP, Spectrum™ Chemical
SDP

Catalog No. 18603414 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-414 50 kg Amber Glass Bottle
18-603-410 125 g Amber Glass Bottle
18-603-411 500 g Amber Glass Bottle
18-603-412 2.5 kg Amber Glass Bottle
18-603-413 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-414 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. IC11050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

IC110, 8029-68-3

Spectrum™ Chemical Ichthammol, USP , also known as ammonium bituminosulfonate, is used to treat certain skin diseases like psoriasis and eczema. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8029-68-3

Specifications

CAS Max % 100%
Boiling Point 130°C/266°F
Ignition Residue 0.5%
Loss on Drying 50%
Packaging Amber Glass Bottle
Quantity 50 kg
Grade USP
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Ichthammol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.