Learn More
Enzo Life Sciences ICI-118,551 (10mg). CAS: 72795-01-8
SDP
Supplier:  Enzo Life Sciences BMLAR1040010
Encompass_Preferred
A highly selective β2-adrenergic receptor antagonist. Purity: ≥98% (HPLC). Solubility: Soluble in water (20mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0326