Learn More
Medchemexpress LLC HY-13951 50mg , ICI 118,551 (hydrochloride) CAS:72795-01-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1395150MG
Encompass_Preferred
HY-13951 50mg ICI 118,551 (hydrochloride) CAS: 72795-01-8 ICI 118,551 (hydrochloride) is a highly selective β2 adrenergic receptor antagonist, with Kis of 0.7, 49.5 and 611 nM for β2, β1 and β3 receptors, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0833