Learn More
Cayman Chemical ICI 118551 (hydrochloride), 72795-01-8, 10 mg
SDP
Supplier:  Cayman Chemical 1559110
Encompass_Preferred
A potent, selective antagonist of the β2-adrenergic receptor (Ki = 0.7 nM for the β2 receptor, compared with 49.5 and 611 nM for β1 and β3 receptors, respectively); active in vivo and often used to evaluate the actions of adrenergic receptor agonists
Catalog No. NC0904861
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.