Learn More
Cayman Chemical ICI 118551hydrochlorIde 10mg
SDP
Catalog No. 502968907
Encompass_Preferred
A potent, selective antagonist of the β2-adrenergic receptor (Ki = 0.7 nM for the β2 receptor, compared with 49.5 and 611 nM for β1 and β3 receptors, respectively); active in vivo and often used to evaluate the actions of adrenergic receptor agonists
Catalog No. 50-296-8907 Supplier Cayman Chemical Supplier No. 1559110 MG
A potent, selective antagonist of the β2-adrenergic receptor (Ki = 0.7 nM for the β2 receptor, compared with 49.5 and 611 nM for β1 and β3 receptors, respectively); active in vivo and often used to evaluate the actions of adrenergic receptor agonists