Learn More
Enzo Life Sciences Icilin (50mg). CAS: 36945-98-9
SDP
Catalog No. 502010433
Encompass_Preferred
Cooling agent. Strongly activates TRPM8 (IC50=0.2µM), cold menthol receptor 1 (CMR1, IC50=0.36µM), a nonselective, cold-activated cation channel and TRPA1 (at 10- to 100-fold higher concentration). Induces currents in TRPM8 expressing HEK 293 cells (EC50=0.36mM) more potently than menthol (ALX-420-042) or low temperatures. Activates TRPM8 with variable latency, followed by extensive desensitization provided calcium is present. The effects of icilin can be impacted by both pH and cytosolic calcium levels. Alternative name: AG 3-5, 3,6-Dihydro-1-(2-hydroxyphenyl)-4-(3-nitrophenyl)-2(1H)-pyrimidinone. Purity: ≥97% (TLC). Solubility: Soluble in DMSO (50mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0433 Supplier Enzo Life Sciences Supplier No. BMLCM1260050
Cooling agent. Strongly activates TRPM8 (IC50=0.2µM), cold menthol receptor 1 (CMR1, IC50=0.36µM), a nonselective, cold-activated cation channel and TRPA1 (at 10- to 100-fold higher concentration). Induces currents in TRPM8 expressing HEK 293 cells (EC50=0.36mM) more potently than menthol (ALX-420-042) or low temperatures. Activates TRPM8 with variable latency, followed by extensive desensitization provided calcium is present. The effects of icilin can be impacted by both pH and cytosolic calcium levels. Alternative name: AG 3-5, 3,6-Dihydro-1-(2-hydroxyphenyl)-4-(3-nitrophenyl)-2(1H)-pyrimidinone. Purity: ≥97% (TLC). Solubility: Soluble in DMSO (50mg/ml). Long Term Storage: Ambient.