Learn More
Sino Biological Recombinant Human HIPK3 Protein (His Tag), Active
Catalog No. 502574774
Encompass_Preferred
Recombinant human HIPK3 (163-562) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-4774 Supplier Sino Biological Supplier No. H0513H500
Recombinant human HIPK3 (163-562) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.