Learn More
Aviva Systems Biology Corp ICK Antibody - C-terminal region (ARP63766_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63766_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ICK. It was validated on Western Blot. . Alternate names: ECO, KIAA0936, LCK2, MGC46090, MRKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-077