Learn More
Medchemexpress LLC ICL-CCIC-0019 100mg | 936498-64-5 | 572.46 | C26H44Br2N4 | 100 MG
SDP
Catalog No. 5000227023
Encompass_Preferred
ICL-CCIC-0019 is a small-molecule inhibitor of choline kinase α (CHKα) that induces G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines. It is supplied as a solid for research use and is available in multiple milligram pack sizes.
  • Inhibits choline kinase α (CHKα) and alters cellular metabolism.
  • Induces G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines.
  • High reported purity suitable for biochemical and cellular assays.
  • Molecular weight 572.46 and molecular formula C26H44Br2N4.
  • Available in small milligram-scale quantities for research applications.
Catalog No. 50-002-27023 Supplier Medchemexpress LLC Supplier No. HY115840100MG
ICL-CCIC-0019 is a small-molecule inhibitor of choline kinase α (CHKα) that induces G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines. It is supplied as a solid for research use and is available in multiple milligram pack sizes.
  • Inhibits choline kinase α (CHKα) and alters cellular metabolism.
  • Induces G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines.
  • High reported purity suitable for biochemical and cellular assays.
  • Molecular weight 572.46 and molecular formula C26H44Br2N4.
  • Available in small milligram-scale quantities for research applications.