Learn More
Medchemexpress LLC ICL-CCIC-0019 10mg | 936498-64-5 | 572.46 g/mol | C26H44Br2N4 | 10 MG
SDP
Catalog No. 5000307830
Encompass_Preferred
ICL-CCIC-0019 is a small-molecule inhibitor of choline kinase α (CHKα) used in preclinical cancer research. It decreases phosphocholine levels and induces G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines.
  • Inhibits choline kinase α (CHKα) activity.
  • Induces G1 cell-cycle arrest and apoptosis in cancer cell lines.
  • Confirmed purity by HPLC: 95.38%.
  • Provided as a dibromide pyridinium salt (C26H44Br2N4).
  • Store sealed at -20°C; in solvent, store at -80°C up to 6 months.
Catalog No. 50-003-07830 Supplier Medchemexpress LLC Supplier No. HY11584010MG
ICL-CCIC-0019 is a small-molecule inhibitor of choline kinase α (CHKα) used in preclinical cancer research. It decreases phosphocholine levels and induces G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines.
  • Inhibits choline kinase α (CHKα) activity.
  • Induces G1 cell-cycle arrest and apoptosis in cancer cell lines.
  • Confirmed purity by HPLC: 95.38%.
  • Provided as a dibromide pyridinium salt (C26H44Br2N4).
  • Store sealed at -20°C; in solvent, store at -80°C up to 6 months.