Learn More
Medchemexpress LLC ICL-CCIC-0019 1mg | 936498-64-5 | 572.46 g·mol⁻¹ | C26H44Br2N4 | 1 MG
SDP
Catalog No. 5000227962
Encompass_Preferred
ICL-CCIC-0019 is a small-molecule inhibitor of choline kinase α (CHKα). It reduces cellular phosphocholine levels and has been reported to induce G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines. The compound is supplied as a solid research reagent intended for biochemical and cellular studies of choline metabolism.
  • Small-molecule CHKα inhibitor for biochemical and cellular studies.
  • Reduces cellular phosphocholine and alters choline metabolism.
  • Induces G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell models.
  • Supplied as a powder with high reported purity suitable for research use.
  • Store sealed at -20 °C, away from moisture and light.
Catalog No. 50-002-27962 Supplier Medchemexpress LLC Supplier No. HY1158401MG
ICL-CCIC-0019 is a small-molecule inhibitor of choline kinase α (CHKα). It reduces cellular phosphocholine levels and has been reported to induce G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines. The compound is supplied as a solid research reagent intended for biochemical and cellular studies of choline metabolism.
  • Small-molecule CHKα inhibitor for biochemical and cellular studies.
  • Reduces cellular phosphocholine and alters choline metabolism.
  • Induces G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell models.
  • Supplied as a powder with high reported purity suitable for research use.
  • Store sealed at -20 °C, away from moisture and light.