Learn More
Medchemexpress LLC ICL-CCIC-0019 50mg | 936498-64-5 | 572.46 | C26H44Br2N4 | 50 MG
SDP
Catalog No. 5000226343
Encompass_Preferred
ICL-CCIC-0019 is a small-molecule inhibitor of choline kinase α (CHKα) used for preclinical research. It decreases phosphocholine levels and induces G1 arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines. The compound is supplied with high chemical purity and should be stored sealed, away from moisture and light at -20°C.
  • Inhibits choline kinase α (CHKα) activity in cellular assays.
  • Induces G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines.
  • High chemical purity suitable for preclinical research use.
  • Supplied as a solid; store sealed, away from moisture and light at -20°C.
  • Molecular formula C26H44Br2N4; molecular weight 572.46 g/mol.
Catalog No. 50-002-26343 Supplier Medchemexpress LLC Supplier No. HY11584050MG
Add to Cart
Add to Cart
ICL-CCIC-0019 is a small-molecule inhibitor of choline kinase α (CHKα) used for preclinical research. It decreases phosphocholine levels and induces G1 arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines. The compound is supplied with high chemical purity and should be stored sealed, away from moisture and light at -20°C.
  • Inhibits choline kinase α (CHKα) activity in cellular assays.
  • Induces G1 cell-cycle arrest, endoplasmic reticulum stress, and apoptosis in cancer cell lines.
  • High chemical purity suitable for preclinical research use.
  • Supplied as a solid; store sealed, away from moisture and light at -20°C.
  • Molecular formula C26H44Br2N4; molecular weight 572.46 g/mol.