Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ICL-CCIC-0019 5mg | 936498-64-5 | 572.46 g·mol⁻1 | C26H44Br2N4 | 5 MG
SDP

Catalog No. 5000307659
Encompass_Preferred

ICL-CCIC-0019 is a small-molecule inhibitor of choline kinase α (CHKα) used in research to modulate phospholipid metabolism and induce cell-cycle arrest and apoptosis in cancer cell lines. Supplied as a solid for laboratory research use only.

  • Purity 97.8% by supplier certificate of analysis.
  • Molecular weight 572.46 g·mol⁻1.
  • Chemical formula C26H44Br2N4.
  • Soluble in DMSO: 8.33 mg/mL (approx. 14.55 mM); ultrasonic recommended.
  • Appearance white to off-white solid.
  • Storage sealed, away from moisture and light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Packaged as a 5 MG research vial.

Catalog No. 50-003-07659 Supplier Medchemexpress LLC Supplier No. HY1158405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ICL-CCIC-0019 is a small-molecule inhibitor of choline kinase α (CHKα) used in research to modulate phospholipid metabolism and induce cell-cycle arrest and apoptosis in cancer cell lines. Supplied as a solid for laboratory research use only.

  • Purity 97.8% by supplier certificate of analysis.
  • Molecular weight 572.46 g·mol⁻1.
  • Chemical formula C26H44Br2N4.
  • Soluble in DMSO: 8.33 mg/mL (approx. 14.55 mM); ultrasonic recommended.
  • Appearance white to off-white solid.
  • Storage sealed, away from moisture and light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Packaged as a 5 MG research vial.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.