Learn More
TARGETMOL CHEMICALS INC ICL-CCIC-0019 5MG
SDP
Catalog No. 502580601
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ICL-CCIC-0019 is an inhibitor of choline kinase alpha (ChoK(alpha)). ICL-CCIC-0019 induces G1 arrest endoplasmic reticulum stress and apoptosis of cancer cell lines. purity: 97%
Catalog No. 50-258-0601 Supplier TARGETMOL CHEMICALS INC Supplier No. T275795MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ICL-CCIC-0019 is an inhibitor of choline kinase alpha (ChoK(alpha)). ICL-CCIC-0019 induces G1 arrest endoplasmic reticulum stress and apoptosis of cancer cell lines. purity: 97%