Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iclaprim | 192314-93-5 | MFCD09837803 | 98.9% | 354.40 | C19H22N4O3 | 50 MG
SDP

Catalog No. 5000248226
Encompass_Preferred

Iclaprim is a selective bacterial dihydrofolate reductase inhibitor supplied for research use to investigate antibacterial activity. It inhibits growth of Staphylococcus aureus, including MRSA, with a reported MIC90 of 0.06 μg/mL and is provided as a characterized research reagent.

  • High purity (98.9%).
  • Molecular weight 354.40; formula C19H22N4O3.
  • Demonstrated activity against S. aureus (MIC90 0.06 μg/mL).
  • Soluble in DMSO; solution available at 10 mM in DMSO.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-48226 Supplier Medchemexpress LLC Supplier No. HY10147950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iclaprim is a selective bacterial dihydrofolate reductase inhibitor supplied for research use to investigate antibacterial activity. It inhibits growth of Staphylococcus aureus, including MRSA, with a reported MIC90 of 0.06 μg/mL and is provided as a characterized research reagent.

  • High purity (98.9%).
  • Molecular weight 354.40; formula C19H22N4O3.
  • Demonstrated activity against S. aureus (MIC90 0.06 μg/mL).
  • Soluble in DMSO; solution available at 10 mM in DMSO.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.