Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iclaprim | 192314-93-5 | MFCD09837803 | 98.9% | 354.40 g/mol | C19H22N4O3 | 1 ML
SDP

Catalog No. 5000248112
Encompass_Preferred

Iclaprim is a selective bacterial dihydrofolate reductase inhibitor used in research on Gram-positive organisms. It shows activity against Staphylococcus aureus, including MRSA, with a reported MIC90 of 0.06 μg/mL. The compound is supplied as a high-purity research reagent and is commonly provided as a 10 mM solution in DMSO for in vitro assays.

  • High purity (98.9%).
  • Active against Staphylococcus aureus, including MRSA (MIC90 0.06 μg/mL).
  • Provided as a 10 mM solution in DMSO (1 mL) for in vitro studies.
  • Recommended solvent DMSO; may require warming and sonication to dissolve.
  • Molecular weight 354.40 g/mol; formula C19H22N4O3.
  • Storage conditions available for powder and in-solution forms to preserve stability.

Catalog No. 50-002-48112 Supplier Medchemexpress LLC Supplier No. HY10147910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iclaprim is a selective bacterial dihydrofolate reductase inhibitor used in research on Gram-positive organisms. It shows activity against Staphylococcus aureus, including MRSA, with a reported MIC90 of 0.06 μg/mL. The compound is supplied as a high-purity research reagent and is commonly provided as a 10 mM solution in DMSO for in vitro assays.

  • High purity (98.9%).
  • Active against Staphylococcus aureus, including MRSA (MIC90 0.06 μg/mL).
  • Provided as a 10 mM solution in DMSO (1 mL) for in vitro studies.
  • Recommended solvent DMSO; may require warming and sonication to dissolve.
  • Molecular weight 354.40 g/mol; formula C19H22N4O3.
  • Storage conditions available for powder and in-solution forms to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.